Would you like to log in again?
Protein Carbonyl ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-312-KI01
89156-130EA 989 USD InStock
89156-130
Protein Carbonyl ELISA Kit, Enzo Life Sciences
Assays ELISAs
The Protein carbonyl ELISA kit is an antibody-based detection set
  • Detecting low levels of carbonylated proteins
  • Validated in various sample types
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
  • Assay Time: 4.5 hours
For quantitative determination of carbonylated protein levels in plasma, other body fluids, cell and tissue extracts.
It has higher sensitivity, lower background, is less labour-intensive and handles more samples per day as compared to colorimetric assays.
Caution: Research Use Only
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty