Would you like to log in again?
Microcystins (Adda Specific) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-319-KI01 850-319-KI01
89156-138EA 1010 USD InStock
CA89156-138 89156-138
Microcystins (Adda Specific) ELISA Kit, Enzo Life Sciences
Assays ELISAs
Polyclonal antibody prepared against the Adda moiety binds to microcystins and nodularins, allowing the congener-independent determination of these toxins and many of its congeners, and does not cross-react with other non-related toxins or compounds
  • Convenient – no pre-sample preparation required
  • Rapid – analyze 41 duplicate samples in <2.5 hours
  • Assay Time: 1 hour 90 minutes
No pre-sample preparation required. Enables simultaneous measurement of multiple samples at reasonable costs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty