Would you like to log in again?
HAT Activity Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-326-KI01
89156-146EA 784.32 USD InStock
89156-146
HAT Activity Assay Kit, Enzo Life Sciences
Assays Protein Assays
Histone acetyltransferases (HATs) have been implicated to play a crucial role in various cellular functions, such as gene transcription, differentiation, and proliferation
The kit utilizes active recombinant HAT as a positive control and acetyl-CoA as a cofactor. Acetylation of peptide substrate by active HAT releases the free form of CoA which then serves as an essential coenzyme for producing NADH. NADH can easily be detected spectrophotometrically upon reacting with a soluble tetrazolium dye. The detection can be continuous and suitable for kinetic studies. The kit provides a simple, straightforward protocol for a complete assay.
HAT Activity Colorimetric Assay Kit offers a convenient, nonradioactive system for a rapid and sensitive detection of HAT activity in mammalian samples.
Delivery information: Kit includes 2X HAT Assay Buffer, HAT Substrate I, HAT Substrate II, NADH Generating Enzyme, NE (Nuclear Extract 4 mg/mL), HAT Reconstitution Buffer.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty