Would you like to log in again?
ADMA Direct (Mouse/Rat) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-327-KI01
89156-148EA 1980 USD InStock
89156-148
ADMA Direct (Mouse/Rat) ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative detection of asymmetrical dimethylarginine (ADMA) in mouse or rat EDTA-plasma and serum
Does not cross-react with SDMA, NMMA or L-arginine.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty