Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Myosin Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
BC-1040-S-L005 BC-1040-S-L001
89156-168EA 857 USD InStock
89156-168 89156-166
Anti-Myosin Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2a

  • For Immunohistochemistry, Western Blot

Recognizes the S1/S2 junction of human ventricular myosin heavy chain beta.

Type: Primary
Antigen: MYOSIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty