Would you like to log in again?
BAFF Soluble (mouse), Detection Set, Enzo Life Sciences
Supplier: Enzo Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
54N-013-KI01
89156-202EA 1850 USD InStock
89156-202
BAFF Soluble (mouse), Detection Set, Enzo Life Sciences
Assays Immunological Assays
For the quantitative determination of soluble mouse BAFF from biological fluids (serum and cell culture supernatant).
Delivery information: Kit contains 1 vial standard (lyophilized) (STD), 1 vial coating antibody (COAT) and 1 vial detection antibody (DET).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty