Would you like to log in again?
SCREEN-WELL® Serotonergic Ligand Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2813-0500 BML-2813-0100
76002-328EA 2916.12 USD InStock
76002-328 89156-272
SCREEN-WELL® Serotonergic Ligand Library, Enzo Life Sciences
Cell Culture Supplements and Additives
The Serotonergic Ligands Plate contains 79 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs.
The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Serotonergic Ligands plate is 1 plate of the larger Neurotransmitter Library which contains over 680 ligands.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty