Would you like to log in again?
SCREEN-WELL® Purinergic Ligand Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2820-0500 BML-2820-0100
76002-342EA 1732.19 USD InStock
76002-342 89156-286
SCREEN-WELL® Purinergic Ligand Library, Enzo Life Sciences
Cell Culture Supplements and Additives
The Screen-Well Purinergic and Adenosine Ligands Plate contains 72 CNS receptor ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs.
The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Purinergic and Adenosine Ligands Plate is 1 plate of the larger Neurotransmitter Library which contains over 650 ligands.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty