Would you like to log in again?
SCREEN-WELL® Protease Inhibitor Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2833-0500 BML-2833-0100
89156-296EA 10443.54 USD InStock
89156-296 89156-294
SCREEN-WELL® Protease Inhibitor Library, Enzo Life Sciences
Cell Culture Supplements and Additives
The Screen-Well™Protease Inhibitor Library includes 53 known protease inhibitors on a single 96-well plate. Inhibitors are supplied dissolved in DMSO at 10 mM and aliquoted into deep-well plates at either 100 or 500 microliters per well. The library includes inhibitors to a broad range of important proteases and is a convenient and cost-effective way to purchase a panel of protease inhibitors for chemical genomics, assay development, and other applications. It includes inhibitors of: Calpains, TPPII, Neutrophil Elastase, Cathepsins, DPPIV, Caspases, Granzyme B, gamma-Secretase, Furin, MMPs, ACE, Kallikrein, Proteasome, Aminopeptidase B, and many more.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty