Would you like to log in again?
SCREEN-WELL® Phosphatase Inhibitor Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2834-0500 BML-2834-0100
89156-300EA 5003.4 USD InStock
89156-300 89156-298
SCREEN-WELL® Phosphatase Inhibitor Library, Enzo Life Sciences
Cell Culture Supplements and Additives
The Screen-Well™ Phosphatase Inhibitor Library contains 33 known phosphatase inhibitors of well-defined activity. Inhibitors are supplied dissolved in DMSO at 10 mM and aliquoted into deep-well plates at either 100 µl or 500 µl per well. The library is ideal for chemical genomics, assay development, and as a reference set for secondary screening. Includes inhibitors of these important phosphatases: Calcineurin(PP2B), JSP-1, PRL-1, CD45, PP1, PRL-3, CDC25, PP2A, PTEN, and many more.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty