Would you like to log in again?
MMP Inhibitor Profiling Kit, Fluorometric, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK016-0001
89156-418EA 1370 USD InStock
89156-418
MMP Inhibitor Profiling Kit, Fluorometric, Enzo Life Sciences
Assays Enzyme Assays
A QUANTIZYME® Assay System Complete assay system designed to examine the specificity of inhibitors against a panel of ten matrix metalloproteinase enzymes. The assays are performed in a convenient 96-well microplate format using a sensitive fluorogenic substrate . Included are active MMP enzymes, assay buffer, a prototypic control inhibitor (NNGH3), and a detailed instruction booklet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty