Would you like to log in again?
ALDetect™ Lipid Peroxidation Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK170-0001
89156-446EA 849 USD InStock
89156-446
ALDetect™ Lipid Peroxidation Assay Kit, Enzo Life Sciences
Assays Cellular Assays
The assay is designed to measure either MDA alone (in hydrochloric acid) or MDA in combination with 4-hydroxyalkenals (in methanesulfonic acid). It is based on the reaction of a chromogenic reagent, N-methyl-2-phenylindole with MDA and 4-hydroxyalkenals to yield a stable chromophore with maximal absorbance at 586 nm.
Lipid peroxidation is a well-established mechanism of cellular injury in both plants and animals, and is used as an indicator of oxidative stress in cells and tissues
Polyunsaturated fatty acid peroxides generate malondialdehyde (MDA) and 4-hydroxyalkenals (HAE) upon decomposition. Measurement of malondialdehyde and 4-hydroxyalkenals has been used as an indicator of lipid peroxidation.
Lipid peroxides are unstable and decompose to form a complex series of compounds including reactive aldehydes.
Delivery information: Kit includes Reagent R1: N-methyl-2-phenylindole, in acetonitrile (3x18 mL), Reagent R2: conc. methanesulfonic acid (16.5 mL), MDA Standard: 1,1,3,3-tetramethoxypropane (TMOP) in TRIS-hydrochloric acid (1 mL), Diluent: ferric iron in methanol (30 mL).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty