Would you like to log in again?
Matrix Metalloproteinase-3 (MMP-3) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BML-AK400-0001
89156-450EA 713 USD InStock
89156-450
Matrix Metalloproteinase-3 (MMP-3) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Assays Enzyme Assays
A QuantiZymeTM Assay System
The MMP-3 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-3 (MMP-3, stromelysin-1) using a thiopeptide as a chromogenic substrate. The assays are performed in a convenient 96 well microplate format.
Delivery information: Kit contains MMP-3 Enzyme (human) (recombinant), Substrate, Inhibitor, Assay Buffer and 96-well Microplate.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty