Would you like to log in again?
Matrix Metalloproteinase-12 (MMP-12) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK402-0001
89156-454EA 713 USD InStock
89156-454
Matrix Metalloproteinase-12 (MMP-12) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Assays Enzyme Assays
A QuantiZymeTM Assay System
The MMP-12 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-12 (MMP-12, macrophage elastase, metalloelastase) using a thiopeptide as a chromogenic substrate. The assays are performed in a convenient 96-well plate format. Included are active enzyme, assay buffer, a prototypic control inhibitor (NNGH), and a detailed instruction booklet.
Delivery information: Kit contains MMP-12 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH), Assay Buffer, 96-well Microplate and a detailed instruction booklet.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty