Would you like to log in again?
Matrix Metalloproteinase-1 (MMP-1) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK404-0001
89156-458EA 700 USD InStock
89156-458
Matrix Metalloproteinase-1 (MMP-1) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Assays Enzyme Assays
A QuantiZymeTM Assay System
The MMP-1 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-1 (MMP-1, interstitial collagenase, fibroblast collagenase) using a thiopeptide as a chromogenic substrate.
Delivery information: Kit includes MMP-1 Enzyme (human) (recombinant), Substrate, inhibitor, Assay Buffer and 96-well Microplate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty