Would you like to log in again?
Matrix Metalloproteinase-1 (MMP-1) Fluorometric Drug Discovery Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK405-0001
89156-460EA 713 USD InStock
89156-460
Matrix Metalloproteinase-1 (MMP-1) Fluorometric Drug Discovery Kit, Enzo Life Sciences
Assays Enzyme Assays
A QuantiZymeTM Assay System
The MMP-1 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-1 (MMP-1, interstitial collagenase) using a quenched fluorogenic peptide: OmniMMPTM fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties
Delivery information: Kit contains MMP-1 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty