Would you like to log in again?
FLUOR DE LYS®-Green HDAC2 Fluorometric Drug Discovery Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK512-0001
89156-510EA 944 USD InStock
89156-510
FLUOR DE LYS®-Green HDAC2 Fluorometric Drug Discovery Assay Kit, Enzo Life Sciences
Assays Enzyme Assays
The HDAC2 Fluorescent Activity Assay/Drug Discovery Kit is a complete assay system designed to measure the lysyl deacetylase activity of the recombinant human HDAC2 included in the kit. The kit is ideal for chemical library screening for candidate inhibitors or kinetic assay of the enzyme under varying conditions.
  • Useful for inhibitor screening or characterizing enzyme kinetics, modulator activity and drug discovery
  • Includes optimal substrate selected from a panel of acetylated sites in p53 and histones
The Fluor de Lys®-Green HDAC2 assay is based on the Fluor de Lys®-Green Substrate and Fluor de Lys® Developer combination
Fluor de Lys®-Green is an especially sensitive substrate for HDAC2. Activity can readily be measured with enzyme amounts in the range of 1-10 ng/well (0.36 – 3.6 nM in 50 mL), thus enabling IC50 determinations for high affinity inhibitors. The assay is highly suitable for high-throughput screening, with a Z'-factor of 0.74.
The assay procedure has two steps. First, the Fluor de Lys®-Green Substrate, which comprises an acetylated lysine side chain, is incubated with HDAC2. Deacetylation of the substrate sensitizes the substrate so that, in the second step, treatment with the Fluor de Lys®Developer produces a fluorophore.
Delivery information: Kit includes HDAC2 (Histone Deacetylase 2) (human, recombinant), FLUOR DE LYS®-Green substrate, 20X FLUOR DE LYS® Developer concentrate, Trichostatin A (HDAC Inhibitor), FLUOR DE LYS®-Green Standard, HDAC Assay Buffer II, Half volume black NBS microplate, Half volume white NBS microplate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty