Would you like to log in again?
LSD1 Fluorometric Drug Discovery Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BML-AK544-0001
89156-520EA 500 USD InStock
89156-520
LSD1 Fluorometric Drug Discovery Kit, Enzo Life Sciences
Assays Enzyme Assays
The LSD1 Fluorimetric Drug Discovery Kit provides human recombinant LSD1 and all reagents for measuring its activity in a sensitive, real-time fluorescent assay.
LSD1 catalyzed demethylation of the Histone H3 Dimethyl Lysine-4 Peptide (H3K4Me2 Peptide) produces hydrogen peroxide. A fluorescent signal is generated via the horseradish peroxidase (HRP) catalyzed reaction of the hydrogen peroxide with the CELLestial® Red Peroxidase Substrate. Although fluorescence detection will be more sensitive (Excitation in range of 530-570 nm; Emission ca. 590 nm), the CELLestial® Red peroxidation product may also be detected by absorbance at 563 nm.
Delivery information: Recombinant human LSD1 (50µg, >1000U)Histone H3 Dimethyl Lysine-4 Peptide SubstrateCELLestial® Red Peroxidase SubstrateHorseradish PeroxidaseH2O2 StandardTranylcypromine (LSD1 inhibitor)Black and clear 1/2-vol. 96-well platesDetailed instructions.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty