Would you like to log in again?
FLUOR DE LYS® SIRT1 Fluorometric Drug Discovery Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK555-0001
89156-522EA 926 USD InStock
89156-522
FLUOR DE LYS® SIRT1 Fluorometric Drug Discovery Assay Kit, Enzo Life Sciences
Assays Enzyme Assays
A FLUOR DE LYS® fluorescent assay system
  • Useful for inhibitor screening or characterizing enzyme kinetics
  • Includes optimal substrate selected from a panel of acetylated sites in p53 and histones
The Fluor de Lys® SIRT1 assay is based on the Fluor de Lys® SIRT1 Substrate and Fluor de Lys® Developer II combination. The assay procedure has two steps. First, the Fluor de Lys® SIRT1 Substrate, which contains a peptide comprising amino acids 379-382 of human p53 (Arg-His-Lys-Lys(Ac)). Deacetylation of the substrate sensitizes the substrate so that, in the second step, treatment with the Fluor de Lys®Developer II produces a fluorophore.
The SIRT1 Fluorescent Activity Assay/Drug Discovery Kit is a complete assay system designed to measure the lysyl deacetylase activity of the recombinant human SIRT1 included in the kit.
Delivery information: Kit includes SIRT1 (Sirtuin 1, hSir2SIRT1) (human, recombinant), FLUOR DE LYS® SIRT1 Deacetylase Substrate, 5X FLUOR DE LYS® Developer II Concentrate, NAD (Sirtuin Substrate), Nicotinamide (Sirtuin Inhibitor), Resveratrol (Sirtuin Activator), Suramin sodium (Sirtuin Inhibitor), FLUOR DE LYS® Deacetylated Standard, Sirtuin Assay Buffer, Half volume microplates.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty