Would you like to log in again?
Granzyme B Assay Kit for Drug Discovery, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK711-0001
89156-542EA 772 USD InStock
89156-542
Granzyme B Assay Kit for Drug Discovery, Enzo Life Sciences
Assays Enzyme Assays
The Granzyme B Assay Kit for Drug Discovery is a complete assay system designed to measure the protease activity of granzyme B. It contains a colorimetric substrate (IEPD-pNA) to measure this activity; cleavage of the p-nitroanilide (pNA) group from the colorimetric substrate increases absorption at 405 nm.
The kit is useful to screen for inhibitors of granzyme B, a potential therapeutic target
Granzyme B is a serine protease found in the lymphocyte granules of cytotoxic T-lymphocytes (CTLs) and natural killer (NK) cells. Granzyme B plays a crucial role in the development of acute graft-vs.-host disease. Thus, the inhibition of this enzyme may prove to be an important tool in controlling immune responses.
An inhibitor 3, 4-Dichloroisocoumarin, is also included for use as a control.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty