Would you like to log in again?
Proteasome 20S Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK740-0001
89156-546EA 772 USD InStock
89156-546
Proteasome 20S Assay Kit, Enzo Life Sciences
Assays Protein Assays
A QuantiZyme™ Assay System
The Suc-LLVY-AMC fluorogenic peptide substrate supplied with this kit is a proteasome substrate commonly used for measuring chymotrypsin-like peptidase activity. The detection of proteolytic activity is based on the release of free AMC fluorophore and offers the following advantages: non-radioactive, convenient real-time kinetics of substrate cleavage, and excellent sensitivity. A proteasome inhibitor, epoxomicin, is included as a test inhibitor for screening purposes.
Fluorogenic, non-radioactive assay designed to measure chymotrypsin-like protease activity of purified 20S proteasome. This 96-well assay is useful for screening inhibitors and modulators of the 20S proteasome.
Delivery information: Kit includes Proteasome 20S (human) (purified), Suc-Leu-Leu-Val-Tyr-AMC, Proteasome Assay Buffer, Sodium Dodecyl Sulfate, Epoxomicin, AMC (7-Amino-4-methylcoumarin) Calibration Standard, Half Volume White Microplates.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty