Would you like to log in again?
Calcineurin Phosphatase Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK804-0001
89156-550EA 626.76 USD InStock
89156-550
Calcineurin Phosphatase Assay Kit, Enzo Life Sciences
Assays Protein Assays
This colorimetric kit is designed for inhibitor screening or kinetic assay of recombinant Calcineurin
  • Safe and non-radioactive assay
  • Convenient 1-step detection, no mixing needed
Activity is determined using the RII phosphopeptide substrate, the most efficient and selective peptide known for calcineurin, and detection of free phosphate released is based on the classic malachite green assay.
Delivery information: Kit includes Recombinant calcineurin, RII phosphopeptide substrate, calmodulin, assay buffer, BIOMOL® GREEN detection reagent,
phosphate standard, microplate and detailed instructions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty