Would you like to log in again?
PTP1B Drug Discovery Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK822-0001
89156-558EA 674 USD InStock
89156-558
PTP1B Drug Discovery Kit, Enzo Life Sciences
Assays Enzyme Assays
The PTP1B Tyrosine Phosphatase Drug Discovery Kit is a colorimetric, non-radioactive assay designed to measure the phosphatase activity of purified PTP1B. This 96-well assay is useful for screening inhibitors and modulators of PTP1B activity.
The kit includes human, recombinant PTP1B (residues 1-322; MW=37.4 kDa), expressed in <italic>E
PTP1B (protein tyrosine phosphatase-1B) is a ubiquitous, nontransmembrane protein tyrosine phosphatase, originally identified in human placenta.It is implicated in the negative regulation of insulin receptor signaling, and is a potential therapeutic target for treatment of type 2 diabetes and obesity. The phosphopeptide substrate supplied with this kit ("IR5", Prod. No. BML-P315, pTyr1158 (a.k.a. pTyr1146, mature peptide numbering)) contains sequence from the insulin receptor beta subunit domain that must be autophosphorylated to achieve full receptor kinase activation. This "activation loop" is the target of several protein phosphatase regulators of insulin signaling, including, notably, PTP1B. The PTP1B inhibitor suramin is supplied as a control for inhibitor detection. Suramin is a reversible and competitive inhibitor of PTP1B, with a Ki of 5.5 µM.
Store all components except the microtiter plate at -70°C for the highest stability. The PTP1B enzyme component must be handled particularly carefully in order to retain maximal enzymatic activity. Thaw it quickly in a RT water bath or by rubbing between fingers, then immediately store on an ice bath. The remaining unused enzyme should be 'snap' frozen, e.g. in liquid nitrogen or a dryice/ethanol bath, and stored at -70°C. If only a few assays are to be performed each day, the PTP1B may be divided into several aliquots (best if ≥%10 µl)) to help minimize freeze/thaw cycles.
coli</italic>. The detection of free phosphate released is based on the classic Malachite green assay and offers the advantages of convenient, 1-step detection and excellent sensitivity, without radioactivity.
Ordering information: Quantity: 96 assays
Contents: PTP 1B Enzyme (human recombinant) (Prod. No. BML-SE332-9092)(5 µg; 0.1 µg/µl in 50 mM HEPES, pH 7.2, 1 mM EDTA, 1 mM DTT, 0.05% NP-40, 10% (v/v) glycerol)Storage: -70°C. Avoid freeze/thaw cyclesSubstrate (“IR5” Insulin Receptor ß, residues 1142-1153, pY1146) (Prod. No. BML-P315-9090)(1 mg; Lyophilized solid)Storage: -20 or -70°C. 2X Assay Buffer (Prod. No. BML-KI467)(20 ml; 100 mM MES, pH 6.0, 300 mM NaCl, 2 mM EDTA, 2 mM DTT, 0.1% NP-40 Storage: -70°C Biomol Red™ Concentrated Phosphate Detection Reagent (Prod. No. BML-KI468)(5 ml; Liquid in screw-cap plastic bottle)Storage: 4°C. Long-term at -70°C. Suramin (PTP1B inhibitor) (Prod. No. BML-KI469)(10 mg; Solid)Storage: -70°C Phosphate Standard (Prod. No. BML-KI470)(0.5 ml; 100 µM in 1X Assay Buffer)Storage: -20 or -70°C ½-VOLUME MICROPLATE (Prod. No. BML-KI101)Storage: RT
Delivery information: Kit contains PTP 1B Enzyme, Substrate (“IR5” Insulin Receptor ?, residues 1142-1153, pY1146), Assay Buffer, Biomol Red™ Concentrated Phosphate Detection Reagent, Suramin (PTP1B inhibitor), Phosphate Standard, and Microplate
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty