Would you like to log in again?
Bz-Val-Gly-Arg-AMC
Supplier: Enzo Life Sciences
BW9375-0005
89156-692EA 158.02 USD InStock
89156-692
Bz-Val-Gly-Arg-AMC
Proteins and Peptides
A substrate for the trypsin-like activity of the 20S proteasome.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty