Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CGRP Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
CA1134-0025 CA1134-0100
89156-752EA 499 USD InStock
89156-752 89156-750
Anti-CGRP Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry

Recognizes human, mouse and rat CGRP.

Type: Primary
Antigen: CGRP
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty