Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CGRP Sheep Polyclonal Antibody

Supplier: Enzo Life Sciences
CA1137-0100 CA1137-0025
89156-758EA 154 USD InStock
89156-758 89156-760
Anti-CGRP Sheep Polyclonal Antibody
Antibodies
Calcitonin gene-related peptide (CGRP) is a 37-amino acid disulphide-bridged sensory neuropeptide present in the central and peripheral nervous systems of many animal species. Two forms of CGRP (α and β) have been identified from both rat and human, and the sequence of chicken CGRP has also been reported, showing striking structural similarity to the mammalian forms. CGRP is abundant in perivascular sensory neurons and is known to be a potent vasodilator. Recent work supports the rôle of CGRP in mediating skin vasodilatation and differential mesenteric artery vasorelaxation, as well as serving as an endogenous myocardial protective substance with a possible involvement in the inhibition of human platelet aggregation. CGRP is frequently co-localised with Substance P and immunoelectron microscopic observations have shown that the peptides are co-stored in single secretory granules both in sensory neurons and in thyroid C-cells. There are many hundreds of reports in the scientific literature describing the localisation and co-existence of CGRP with other transmitter molecules in a vast array of mammalian and non-mammalian species.

  • For Immunohistochemistry

Recognizes CGRP in many mammalian species, including rat, rabbit and human.

Type: Primary
Antigen: CGRP
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty