Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Bromo-cADP-ribose ≥95%

Supplier: Enzo Life Sciences
Warning

BML-CA417-0100 BML-CA417-0500
89156-868EA 291.85 USD InStock
89156-868 89156-866
8-Bromo-cADP-ribose ≥95%
8-Bromo-cADP-ribose
cADPR antagonist.

cADPR antagonist. Blocks the cADPR-induced release of Ca²⁺ from sea urchin egg homogenates at 1 - 3.5 µM.

Soluble in water (5 mg/mL).


Formula: C₁₅H₂₀BrN₅O₁₃P₂ 
Storage Temperature: Freezer
CAS Number: 151898-26-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/34/92/45403492.jpg
Order Now
Specification Test Results
Purity ≥95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty