Would you like to log in again?
2',5'-Dideoxyadenosine ≥98% (by TLC)
Supplier: Enzo Life Sciences
Synonyms: (2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-methyltetrahydrofuran-3-ol
BML-CN110-0005 BML-CN110-0025
89156-996EA 160.19 USD InStock
89156-996 89156-998
2',5'-Dideoxyadenosine ≥98% (by TLC)
2',5'-Dideoxyadenosine
Adenylate cyclase inhibitor
A useful, cell-permeable inhibitor of adenylate cyclase (ID50=250µM). Inhibits growth and caues cell differentiation (1.5µM) in K562 cells.
https://us-legacy.vwr.com/stibo/web/std.lang.all/35/37/45403537.jpg
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty