Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Met Enkephalin Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
EA1150-0100 EA1150-0025
89157-436EA 154 USD InStock
89157-436 89157-440
Anti-Met Enkephalin Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry

Recognizes methionine enkephalin in a wide range of species.

Type: Primary
Antigen: MET ENKEPHALIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Species Independent

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty