Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride ≥98% (by UHPLC)

Supplier: Enzo Life Sciences
Synonyms: Terbinafine Hydrochloride(E)-N,6,6-Trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine hydrochloride

BML-EI318-0100
89158-060EA 100.61 USD InStock
89158-060
trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride ≥98% (by UHPLC)
trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride
Inhibits ergosterol biosynthesis at the level of squalene epoxidase. Potent non-competitive inhibitor of squalene epoxidase from Candida (Ki=30 nM), inhibiting mammalian enzymes at higher concentrations (4-77 µM). It inhibits HUVEC proliferation and angiogenesis and displays interesting anti-inflammatory and free radical scavenging activities. Clinically useful antifungal agent.

  • Squalene epoxidase inhibitor

White to off-white solid.

Soluble in ethanol (45 mg/ml), DMSO (30 mg/ml) or water (3 mg/ml).


Formula: C₂₁H₂₅N
MW: 327.90 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00145430
CAS Number: 78628-80-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/37/67/45403767.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty