Would you like to log in again?
Mg2+/ATP Activating Solution
Supplier: Enzo Life Sciences
EW9805-0100
89158-298EA 61.72 USD InStock
89158-298
Mg2+/ATP Activating Solution
Enzymes
Liquid. In 50mM TRIS-Cl, pH 7.5, containing 0.1M ATP and 0.1M MgCl2.
ubiquitin thioester conjugation, proteasomal degradation, SUMO conjugation).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty