Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FN1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
FG6020-0100
89158-460EA 273 USD InStock
89158-460
Anti-FN1 Mouse Monoclonal Antibody
Antibodies
Fibronectins are high molecular weight, disulphide-linked, dimeric cell adhesion glycoproteins found in basement membranes and in the interstitial connective tissue matrix. A single fibronectin gene is subject to alternative splicing in a cell-type-, development- and age-regulated manner which gives rise to multiple molecular forms. In addition to their prominent role in adhesion, fibronectins have been reported to mediate various aspects of cellular interaction, including migration during development and wound-healing, haemostasis, and the regulation of cell growth and differentiation. Cellular fibronectins (cFn) are found in low amounts in normal human plasma and tissues, but they are abundant in the plasma of carcinoma patients and in the stroma of various carcinomas. In contrast, a soluble form of fibronectin produced by hepatocytes is readily detectable in plasma and becomes deposited in pericellular matrices and within tissues. This form of fibronectin, referred to as ‘plasma fibronectin’ (pFn), differs from cFn by the absence of an amino acid sequence, known as extra domain A1.

  • For Immunocytochemistry, Western Blot

Recognizes the cell binding region of fibronectin. Detects a band of ~225-250kDa by Western blot.

Type: Primary
Antigen: FIBRONECTIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty