Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SPTAN1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
FG6090-0100 FG6090-0500
89158-462EA 309 USD InStock
89158-462 89158-464
Anti-SPTAN1 Mouse Monoclonal Antibody
Antibodies
Fodrin, also referred to as non-erythroid (αII-; brain) spectrin, is a tetrameric (αγ)2 actin-binding, fibrous protein, widely distributed in vertebrates, which forms part of the sub-membranous cytoskeleton within many cell types including neurons, and is particularly abundant with axons. The α-subunits of fodrins and spectrin are highly conserved phylogenetically, with the exception of human α-fodrin, which shares only 55-59% homology with erythroid-specific α-spectrins. The β-subunits of spectrin (and γ-subunits of fodrins) are species specific. The interleukin-1 converting enzyme (ICE) family of proteases has been implicated as important effectors of the apoptotic pathway, perhaps acting hierarchically in a protease cascade. Neuronal fodrin is known to be cleaved by calpain following ischaemic insult and it has been proposed that calpain and an unidentified protease play a role in the onset of neuronal death following transient forebrain ischaemia. Recently, an ICE-like protease has been implicated in the early cleavage of fodrin, producing a 150kDa fragment, proximal to CPP32 in Fas-induced and C2-ceramide mediated apoptosis. A cleavage product of α-fodrin has been proposed as a candidate autoantigen in primary Sjögren’s syndrome and α-fodrin has been shown to be the source of a so-called ‘inhibitory protein factor’ family, members of which have been shown to inhibit both GABA and ATP-dependent glutamate uptake into purified synaptic vesicles.

  • For Immunohistochemistry, Western Blot

Recognizes mammalian and chicken alpha-fodrin

Type: Primary
Antigen: SPTAN1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mammals

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty