Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Gastrin Releasing Peptide Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
GA1166-0100 GA1166-0025
89158-672EA 155 USD InStock
89158-672 89158-674
Anti-Gastrin Releasing Peptide Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry

Recognizes GRP in a wide range of species.

Type: Primary
Antigen: GASTRIN RELEASING PEPTIDE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Species Independent

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty