Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GAD1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
GC3008-0100 GC3008-0025
89158-704EA 499 USD InStock
89158-704 89158-702
Anti-GAD1 Rabbit Polyclonal Antibody
Antibodies
Glutamic acid decarboxylase (GAD; E.C. 4.1.1.15) is the enzyme responsible for the conversion of glutamic acid to γ-aminobutyric acid (GABA) - the major inhibitory transmitter in higher brain regions. Two molecular forms of GAD (GAD65 and GAD67) are known from rat, cat, pig and man, and both forms are expressed in the CNS and in pancreatic islet endocrine cells. A recent Western blotting study has identified that the isoforms are regionally distributed in the brains of rats and mice. In rat pancreas, GAD65 and GAD67 appear to be differentially localised, GAD65 primarily in insulin-containing (β) cells and GAD67 in glucagon-containing (A) cells. Evidence from knock-out mice suggests that GABA generated by pancreatic GAD isoforms is not critically involved in normal islet cell development and morphology. However, β-cell-specific GAD expression has been shown to be required for the development of autoimmune diabetes in non-obese diabetic (NOD) mice.

  • For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Recognizes human, mouse, rat and cat GAD. Detects two isoforms of ~65kDa and ~67 kDa by Western Blot.

Type: Primary
Antigen: GLUTAMATE DECARBOXYLASE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty