Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GAD1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
GC3208-0100 GC3208-0025
89158-708EA 529 USD InStock
89158-708 89158-706
Anti-GAD1 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Western Blot

Recognizes an N-terminal epitope in mouse, rat, monkey and human tissues. The antibody consistently labels neuronal GAD in the CNS and GAD-containing cells in mammalian (not mouse) pancreas.

Type: Primary
Antigen: GLUTAMATE DECARBOXYLASE
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty