Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxyphenylretinamide ≥97% (by TLC)

Supplier: Enzo Life Sciences
Danger
Synonyms: (2E,4E,6E,8E)-N-(4-Hydroxyphenyl)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenamideFenretinide4-HPRall-trans-N-(4-Hydroxyphenyl)retinamideN-(4-Hydroxyphenyl)-all- trans retinamideRetinoic Acid p-Hydroxyanilide

BML-GR103-0005
89158-744EA 150.09 USD InStock
89158-744
4-Hydroxyphenylretinamide ≥97% (by TLC)
4-Hydroxyphenylretinamide
Retinoic acid receptor ligand.

An analog of all trans retinoic acid that shows antiproliferative activity on cultured human breast cancer cells. Highly selective activator of retinoid receptors. Induces apoptosis in malignant hemopoietic cell lines.

Soluble in DMSO (25 mg/mL) or 100% ethanol (25 mg/mL).


Formula: C₂₆H₃₃NO₂
MW: 391.56 g/mol
MDL Number: MFCD00792674
CAS Number: 65646-68-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/39/03/45403903.jpg
Order Now
Specification Test Results
Source Synthetic
Purity ≥97% (TLC)
Appearance Orange solid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty