Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HMOX1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
HC3001-0025 HC3001-0100
89160-052EA 151 USD InStock
89160-052 89160-054
Anti-HMOX1 Rabbit Polyclonal Antibody
Antibodies
The rate limiting enzyme in heme catabolism, heme oxygenase, was first described in 1968 and was thought to be a species of cytochrome P450. However, subsequent work showed the enzyme to be a protein distinct from the P450 family and capable of catalysing the degradation of heme to biliverdin and iron, with the concomitant release of carbon monoxide. This enzyme activity is highly conserved throughout the Animal Kingdom and has been identified in algae and plants as well as in all higher species. Heme oxygenase-1 (HO-1) was first purified to homogeneity from rat liver and pig spleen and shown to have a molecular weight of 32kDa. Increased expression of HO-1 can be induced by a large number of structurally unrelated pharmacological and other agents, including cytokines and oxidants, as well as by heat shock and other forms of cellular stress. Baseline levels of HO-1 expression are high in certain tissues, including spleen and liver, but enriched enzyme preparations are recommended for experimental analysis. Recent data have demonstrated a cytoprotective role for HO-1, preventing cell death by modulating intracellular iron levels.

  • For Immunohistochemistry, Western Blot

Recognizes human heme oxygenase 1.

Type: Primary
Antigen: HMOX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty