Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD41 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
IG6065-0100
89160-146EA 273 USD InStock
89160-146
Anti-CD41 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Western Blot

Recognizes human alphaIIb integrin.

Type: Primary
Antigen: CD41
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty