Would you like to log in again?
p-Nitroaniline Standard, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-KI106-0001
89160-286EA 49.33 USD InStock
89160-286
p-Nitroaniline Standard, Enzo Life Sciences
Standards Organic Standards
pNA Calibration Standard (p-nitroaniline).  Chromogenic chromophore (λmax=400nm). Reference compound.
Formulation: 0.3mM (0.2mg/ml) in 50mM HEPES, pH 7.4, 100mM sodium chloride, 0.1% CHAPys, 10mM dithiothreitol, 1mM EDTA, 10% glycerol.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty