Would you like to log in again?
AMC (7-Amino-4-methylcoumarin) Standard, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-KI107-0001
89160-288EA 50.46 USD InStock
89160-288
AMC (7-Amino-4-methylcoumarin) Standard, Enzo Life Sciences
Protein Linking and Labeling Systems
Fluorogenic chromophore. Ex(max): 380nm, Em(max): 460nm. Reference compound.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty