Would you like to log in again?
Nuclear extract from HeLa cells, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-KI140-0100
89160-320EA 150.08 USD InStock
89160-320
Nuclear extract from HeLa cells, Enzo Life Sciences
Cells
Prepared by high salt extraction of HeLa nuclei (human cervical cancer cell line), this extract is a rich source of HDAC activity, useful for inhibitor screening or immunoprecipitation of HDAC containing complexes. Typically, 0.5 µl or less per well is enough for assay with the HDAC FLUOR DE LYS® fluorometric activity assay.
Formulation: Liquid in 0.1 M KCl, 20 mM HEPES/NaOH, pH 7.9, 20% (v/v) glycerol, 0.2 mM EDTA, 0.5 mM DTT, 0.5 mM PMSF.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty