Would you like to log in again?
FLUOR DE LYS® Developer II Concentrate, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-KI176-1250
89160-344EA 294.2 USD InStock
89160-344
FLUOR DE LYS® Developer II Concentrate, Enzo Life Sciences
Protein Extraction Protein Extraction Reagents
FLUOR DE LYS® Developer II is an essential reagent for signal generation in assays done with the FLUOR DE LYS®-HDAC8, FLUOR DE LYS®-SIRT1 and FLUOR DE LYS®-SIRT2 substrates.
Deacetylation of the FLUOR DE LYS® substrate sensitizes it so that, in a second step, treatment with the FLUOR DE LYS® Developer II produces a fluorophore. Typically, an 'inhibitor stop' will be included in the 1x Developer dilution (e.g. nicotinamide for sirtuins).
Ordering information: Quantity: NOTE: This product is supplied packaged in 5 tubes (250 µl per tube) for a total volume of 1.25 ml. This packaging is used for optimal stability of the developer.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty