Would you like to log in again?
Kemptide
Supplier: Enzo Life Sciences
BML-P107-0001
89160-932EA 70.53 USD InStock
89160-932
Kemptide
Proteins and Peptides
PKA substrate
Phosphate acceptor peptide. Substrate for Ser/Thr protein kinases. Km=4.7µM for cAMP-dependent protein kinase (PKA). The sequence defines the phosphorylation site of porcine liver pyruvate kinase.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty