Would you like to log in again?
GP-AMC, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-P189-0005
89161-046EA 125.63 USD InStock
89161-046
GP-AMC, Enzo Life Sciences
Proteins and Peptides
DPP substrate
Fluorogenic substrate for dipeptidyl peptidase IV (DPIV, DPPIV, CD26) (kcat/Km=2.6 x 105s-1M-1), DPPII/VII, DPP8 and DPP9, FAP (fibroblast activation protein), and likely POP/PREP, but not glutamate carboxypeptidase II. Can be used in vitro, for inhibitor screening, and in biological fluids. Ex = 380 nm, Em=460 nm.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty