Would you like to log in again?
Protein Kinase C (20-28) (Myristoylated)
Supplier: Enzo Life Sciences
BML-P205-0500
89161-074EA 167.48 USD InStock
89161-074
Protein Kinase C (20-28) (Myristoylated)
Enzymes
PKC inhibitor
Pseudosubstrate sequence from PKCalpha and ß. Highly specific inhibitor, which is N-terminal myristoylated to allow membrane permeability. IC50=8 µM for TPA activation of MARCKS phosphorylation in fibroblast primary cultures, and 98% inhibition at 100 µM. Partially reverses multidrug resistance in human breast cancer cells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty