Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PBF1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW0435-0100 PW0435-0025
89161-838EA 440.51 USD InStock
89161-838 89161-836
Anti-PBF1 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes Arabidopsis proteasome 20S (PBF1/beta6 subunit).

Type: Primary
Antigen: PBF1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Arabidopsis thaliana

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty