Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-E6AP Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW0535-0025 PW0535-0100
89161-896EA 169 USD InStock
89161-896 89161-898
Anti-E6AP Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Western Blot

Recognizes human E6AP.

Type: Primary
Antigen: E6AP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty