Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BLM10 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

PW0570-0025
89161-904EA 200.93 USD InStock
89161-904
Anti-BLM10 Rabbit Polyclonal Antibody
Antibodies
Blm10 (mammalian homolog PA200) is a large 246kDa regulatory protein that binds to the 20S proteasome core particle (CP). Blm10 recognizes pre-activated open gate CP which is assumed to exist in equilibrium with inactive closed gate CP. Blm10 distinguishes gate conformations, regulates CP activation, and has been shown to be involved in CP maturation.

  • For Western Blot

Recognizes Saccharomyces cervisiae Blm10.

Type: Primary
Antigen: BLM10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Saccharomyces Cerevisiae

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty