Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-USP21 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW0585-0025 PW0585-0100
89161-914EA 156 USD InStock
89161-914 89161-916
Anti-USP21 Rabbit Polyclonal Antibody
Antibodies
USP21 is a ubiquitin-specific protease that catalyzes the hydrolysis of ubH2A thereby acting to relieve ubH2A-specific repression. USP21 is also thought to be involved with hepatocyte regeneration. Furthermore, in addition to its deubiquitinylating activity, USP21 is capable of removing NEDD8 from NEDD8 conjugates but has no effect on SUMO conjugates.

  • For Western Blot

Recognizes human USP21.

Type: Primary
Antigen: USP21
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty